Product Details

SNP ID
rs200743394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:54255285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGAACTTAAACTTGCAGCCTGT[C/T]CTAGGGTGTCTGAGCTCCTTCACTT
Phenotype
MIM: 102595
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACYP2 PubMed Links

Gene Details

Gene
ACYP2
Gene Name
acylphosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320586.1 988 Intron NP_001307515.1
NM_001320587.1 988 Intron NP_001307516.1
NM_001320588.1 988 Intron NP_001307517.1
NM_001320589.1 988 Intron NP_001307518.1
NM_001320590.1 988 Intron NP_001307519.1
NM_138448.3 988 Intron NP_612457.1
XM_017005406.1 988 Intron XP_016860895.1
XM_017005407.1 988 Intron XP_016860896.1
XM_017005408.1 988 Intron XP_016860897.1
XM_017005409.1 988 Intron XP_016860898.1
XM_017005410.1 988 Intron XP_016860899.1
XM_017005411.1 988 Intron XP_016860900.1
XM_017005412.1 988 Intron XP_016860901.1
XM_017005413.1 988 Intron XP_016860902.1
Gene
TSPYL6
Gene Name
TSPY like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003937.2 988 Silent Mutation AGA,AGG R289R NP_001003937.2

View Full Product Details