Product Details

SNP ID
rs200812680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:134456738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCACTCAAACATCTCGTAGTGA[C/T]GTGTCTGCCTCACCCTCGGCACCAT
Phenotype
MIM: 601774
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGAT5 PubMed Links

Gene Details

Gene
MGAT5
Gene Name
mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase
There are no transcripts associated with this gene.

Gene
TMEM163
Gene Name
transmembrane protein 163
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030923.4 1020 Missense Mutation CAT,CGT H283R NP_112185.1
XM_011511950.2 1020 Missense Mutation CAT,CGT H172R XP_011510252.1
XM_017005060.1 1020 Intron XP_016860549.1
XM_017005061.1 1020 Missense Mutation CAT,CGT H172R XP_016860550.1
XM_017005062.1 1020 Intron XP_016860551.1

View Full Product Details