Product Details

SNP ID
rs200868239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:161995301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTCTAATTAACTGAAACTCACC[A/G]TCTGCTGTTCCATGAATAAGGAGGT
Phenotype
MIM: 102720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPP4 PubMed Links

Gene Details

Gene
DPP4
Gene Name
dipeptidyl peptidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001935.3 2685 Silent Mutation GAC,GAT D708D NP_001926.2
XM_005246371.2 2685 Silent Mutation GAC,GAT D707D XP_005246428.1

View Full Product Details