Product Details

SNP ID
rs200895740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:179950598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCTATCACTGCTGTCAGAATCA[C/G]AGTCGGATGAGTCAGACTCTGAAGA
Phenotype
MIM: 615186
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CWC22 PubMed Links

Gene Details

Gene
CWC22
Gene Name
CWC22 homolog, spliceosome-associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020943.2 2354 Missense Mutation TCT,TGT S685C NP_065994.1
XM_005246726.2 2354 Missense Mutation TCT,TGT S685C XP_005246783.1

View Full Product Details