Product Details

SNP ID
rs200965294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:8682467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGGCAGAACCAGGCGTCCAGGA[C/T]GCCGCTGACCACCCTCAACACGGAT
Phenotype
MIM: 600386
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ID2 PubMed Links

Gene Details

Gene
ID2
Gene Name
inhibitor of DNA binding 2, HLH protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002166.4 Intron NP_002157.2
Gene
ID2-AS1
Gene Name
ID2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details