Product Details

SNP ID
rs200993212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:186590458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTAGACGTGGACAGTCCTGCCGA[C/G]TACTCTGGCCCCGAGGGAAGTTACT
Phenotype
MIM: 193210
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITGAV PubMed Links

Gene Details

Gene
ITGAV
Gene Name
integrin subunit alpha V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144999.2 396 Intron NP_001138471.1
NM_001145000.2 396 Missense Mutation GAC,GAG D40E NP_001138472.1
NM_002210.4 396 Missense Mutation GAC,GAG D40E NP_002201.1

View Full Product Details