Product Details
- SNP ID
-
rs199500112
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:88173364 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCTCTGAGCTCCTTCCAAAAGAT[C/G]AATTAAATGGTGAGTGCTCCCACCT
- Phenotype
-
MIM: 611261
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
THNSL2
PubMed Links
Gene Details
- Gene
- THNSL2
- Gene Name
- threonine synthase like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001244676.1 |
2127 |
Missense Mutation |
CAA,GAA |
Q72E |
NP_001231605.1 |
NM_001244678.1 |
2127 |
Intron |
|
|
NP_001231607.1 |
NM_018271.4 |
2127 |
Missense Mutation |
CAA,GAA |
Q72E |
NP_060741.3 |
XM_005264400.4 |
2127 |
Missense Mutation |
CAA,GAA |
Q72E |
XP_005264457.1 |
XM_005264401.4 |
2127 |
Missense Mutation |
CAA,GAA |
Q72E |
XP_005264458.1 |
XM_005264402.3 |
2127 |
Missense Mutation |
CAA,GAA |
Q72E |
XP_005264459.1 |
XM_005264403.4 |
2127 |
Missense Mutation |
CAA,GAA |
Q72E |
XP_005264460.1 |
XM_005264405.4 |
2127 |
Missense Mutation |
CAA,GAA |
Q72E |
XP_005264462.1 |
XM_006712041.3 |
2127 |
Missense Mutation |
CAA,GAA |
Q72E |
XP_006712104.1 |
XM_006712042.3 |
2127 |
Missense Mutation |
CAA,GAA |
Q72E |
XP_006712105.1 |
XM_006712043.2 |
2127 |
Missense Mutation |
CAA,GAA |
Q72E |
XP_006712106.1 |
XM_006712044.3 |
2127 |
Missense Mutation |
CAA,GAA |
Q72E |
XP_006712107.1 |
XM_011532953.2 |
2127 |
Missense Mutation |
CAA,GAA |
Q72E |
XP_011531255.1 |
XM_011532954.2 |
2127 |
Missense Mutation |
CAA,GAA |
Q72E |
XP_011531256.1 |
XM_011532955.2 |
2127 |
Intron |
|
|
XP_011531257.1 |
View Full Product Details