Product Details

SNP ID
rs199611703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73385902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGATCTGCCATGGCCGGGCGAG[C/G]TGGAGGAGGAGGAGGAGGAGGAGGA
Phenotype
MIM: 606844
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALMS1 PubMed Links
Additional Information
For this assay, SNP(s) [rs72319667] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ALMS1
Gene Name
ALMS1, centrosome and basal body associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015120.4 145 Missense Mutation CTG,GTG L12V NP_055935.4

View Full Product Details