Product Details

SNP ID
rs199533069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73766461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGGGTCTGGGTGTGGAAATGTC[A/G]AGGAGCTGAGTGACCCTGGATCTGA
Phenotype
MIM: 603092
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP11 PubMed Links

Gene Details

Gene
DUSP11
Gene Name
dual specificity phosphatase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003584.2 934 Nonsense Mutation CGA,TGA R298* NP_003575.2

View Full Product Details