Product Details

SNP ID
rs199535487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:136115417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTGACGTTGGCAAAGATGAAGT[C/T]GGGAATAGTCAGCAGGAGGGCAGGG
Phenotype
MIM: 162643
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXCR4 PubMed Links

Gene Details

Gene
CXCR4
Gene Name
C-X-C motif chemokine receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008540.1 827 Missense Mutation AAC,GAC N175D NP_001008540.1
NM_003467.2 827 Missense Mutation AAC,GAC N171D NP_003458.1

View Full Product Details