Product Details

SNP ID
rs200527608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233617997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTCATGGATTTCGCCGATGCTC[A/G]ATGGAAAGCACAAGTACGAAGTTTG
Phenotype
MIM: 606433
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UGT1A8 PubMed Links

Gene Details

Gene
UGT1A8
Gene Name
UDP glucuronosyltransferase family 1 member A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019076.4 353 Missense Mutation CAA,CGA Q97R NP_061949.3

View Full Product Details