Product Details

SNP ID
rs200528447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202884526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCCATGTGGCCAGAGAGGGTCA[C/T]TATGGGAGTCTAGAAAGGAAGAACC
Phenotype
MIM: 616620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR12 PubMed Links

Gene Details

Gene
WDR12
Gene Name
WD repeat domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018256.3 1378 Missense Mutation ATG,GTG M251V NP_060726.3
XM_011511469.2 1378 Missense Mutation ATG,GTG M113V XP_011509771.1

View Full Product Details