Product Details
- SNP ID
-
rs200051569
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:151416626 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTAAGAATTCAGACAAAAATGTA[C/T]GTACTAGAGCACTTTGGGTGATATC
- Phenotype
-
MIM: 608952
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RIF1
PubMed Links
Gene Details
- Gene
- RIF1
- Gene Name
- replication timing regulatory factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001177663.1 |
387 |
Missense Mutation |
CGT,TGT |
R116C |
NP_001171134.1 |
NM_001177664.1 |
387 |
Missense Mutation |
CGT,TGT |
R116C |
NP_001171135.1 |
NM_001177665.1 |
387 |
Missense Mutation |
CGT,TGT |
R116C |
NP_001171136.1 |
NM_018151.4 |
387 |
Missense Mutation |
CGT,TGT |
R116C |
NP_060621.3 |
XM_005246665.3 |
387 |
Missense Mutation |
CGT,TGT |
R116C |
XP_005246722.1 |
XM_011511395.2 |
387 |
Missense Mutation |
CGT,TGT |
R116C |
XP_011509697.1 |
XM_017004422.1 |
387 |
Missense Mutation |
CGT,TGT |
R116C |
XP_016859911.1 |
XM_017004423.1 |
387 |
Missense Mutation |
CGT,TGT |
R116C |
XP_016859912.1 |
XM_017004424.1 |
387 |
Missense Mutation |
CGT,TGT |
R80C |
XP_016859913.1 |
XM_017004425.1 |
387 |
Missense Mutation |
CGT,TGT |
R116C |
XP_016859914.1 |
View Full Product Details