Product Details

SNP ID
rs200402344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24037791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAAGACACCGTTGCATTTCAAC[C/G]AGGTTCCTAGATAAAATTTCCACTT
Phenotype
MIM: 600620 MIM: 616234
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FKBP1B PubMed Links

Gene Details

Gene
FKBP1B
Gene Name
FK506 binding protein 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322963.1 1835 Intron NP_001309892.1
NM_001322964.1 1835 Intron NP_001309893.1
NM_004116.4 1835 Intron NP_004107.1
NM_054033.3 1835 Intron NP_473374.1
XM_011532699.2 1835 Intron XP_011531001.1
XM_017003593.1 1835 Intron XP_016859082.1
XM_017003594.1 1835 Intron XP_016859083.1
XM_017003595.1 1835 Intron XP_016859084.1
XM_017003596.1 1835 Intron XP_016859085.1
Gene
WDCP
Gene Name
WD repeat and coiled coil containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142319.1 1835 Silent Mutation CTC,CTG L568L NP_001135791.1
NM_025203.2 1835 Silent Mutation CTC,CTG L568L NP_079479.1
XM_017005029.1 1835 Silent Mutation CTC,CTG L568L XP_016860518.1

View Full Product Details