Product Details

SNP ID
rs199871119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176116992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATGGAATGCAAACCTGTGGACT[G/T]CTCCCGTCTCTGGCCAAAAGAGAAG
Phenotype
MIM: 142984 MIM: 142986 MIM: 142982
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HOXD10 PubMed Links

Gene Details

Gene
HOXD10
Gene Name
homeobox D10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002148.3 229 Silent Mutation CTG,CTT L53L NP_002139.2
Gene
HOXD11
Gene Name
homeobox D11
There are no transcripts associated with this gene.

Gene
HOXD9
Gene Name
homeobox D9
There are no transcripts associated with this gene.

Gene
LOC107985805
Gene Name
sine oculis-binding protein homolog
There are no transcripts associated with this gene.

View Full Product Details