Product Details

SNP ID
rs200165996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88526462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACATGGTCGACTATCAGCCCTAC[A/T]GGAAGCACAAATACTCCAGGGTCAC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TEX37 PubMed Links

Gene Details

Gene
TEX37
Gene Name
testis expressed 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152670.2 202 Missense Mutation AGG,TGG R29W NP_689883.1
XM_005264188.3 202 Missense Mutation AGG,TGG R29W XP_005264245.3
XM_011532691.1 202 Missense Mutation AGG,TGG R29W XP_011530993.1
XM_011532692.1 202 Missense Mutation AGG,TGG R29W XP_011530994.1

View Full Product Details