Product Details
- SNP ID
-
rs200501578
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:191836626 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACGGTGTGCAGGGTTTCCTCCAGG[A/G]ATTTGTTTTCATCCGGAAGCTCCTC
- Phenotype
-
MIM: 606728
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SDPR
PubMed Links
Gene Details
- Gene
- SDPR
- Gene Name
- serum deprivation response
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004657.5 |
930 |
Missense Mutation |
TCC,TTC |
S192F |
NP_004648.1 |
View Full Product Details