Product Details

SNP ID
rs199817907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:33585015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTATTGTCCTGCTGGTAGCCAC[C/T]ACCCTGGTATCCACTTCCTGTATAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM98A PubMed Links

Gene Details

Gene
FAM98A
Gene Name
family with sequence similarity 98 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304538.1 1459 Missense Mutation AGT,GGT S245G NP_001291467.1
NM_015475.4 1459 Missense Mutation AGT,GGT S440G NP_056290.3

View Full Product Details