Product Details

SNP ID
rs199972115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:121728299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAACCATTTTACCTGGCTATCCA[C/T]AGTCTTCTCAGGAGTGTCAAGAGTA
Phenotype
MIM: 611970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NIFK PubMed Links

Gene Details

Gene
NIFK
Gene Name
nucleolar protein interacting with the FHA domain of MKI67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032390.4 759 Missense Mutation ATG,GTG M228V NP_115766.3
Gene
NIFK-AS1
Gene Name
NIFK antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details