Product Details

SNP ID
rs199903182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202884467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGTCCCAAGATGCACTGCAGAT[C/T]TCTTCAGCATCTGACCACAGAACTG
Phenotype
MIM: 616620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR12 PubMed Links

Gene Details

Gene
WDR12
Gene Name
WD repeat domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018256.3 1437 Silent Mutation GAA,GAG E270E NP_060726.3
XM_011511469.2 1437 Silent Mutation GAA,GAG E132E XP_011509771.1

View Full Product Details