Product Details

SNP ID
rs200056347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:160275690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGGTCATTAGACGTCTCGACAG[C/T]CACCTGCTGTTGACCACTTGCCTCC
Phenotype
MIM: 602310
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100505984 PubMed Links

Gene Details

Gene
LOC100505984
Gene Name
uncharacterized LOC100505984
There are no transcripts associated with this gene.

Gene
RBMS1
Gene Name
RNA binding motif single stranded interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002897.4 1576 Missense Mutation ACT,GCT T387A NP_002888.1
NM_016836.3 1576 Missense Mutation ACT,GCT T390A NP_058520.1
XM_005246737.1 1576 Missense Mutation ACT,GCT T406A XP_005246794.1
XM_005246738.2 1576 Missense Mutation ACT,GCT T405A XP_005246795.1
XM_005246739.2 1576 Missense Mutation ACT,GCT T404A XP_005246796.1
XM_005246740.1 1576 Missense Mutation ACT,GCT T403A XP_005246797.1
XM_005246741.3 1576 Missense Mutation ACT,GCT T403A XP_005246798.1
XM_017004626.1 1576 Missense Mutation ACT,GCT T386A XP_016860115.1

View Full Product Details