Product Details
- SNP ID
-
rs199988182
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:170716427 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCTCCAGGCTCTTCCACCCGTGG[A/T]CCGCCGACATGCCGGCGCACTCGCC
- Phenotype
-
MIM: 609391
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC01124
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3749036] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC01124
- Gene Name
- long intergenic non-protein coding RNA 1124
There are no transcripts associated with this gene.
- Gene
- LOC101926913
- Gene Name
- uncharacterized LOC101926913
There are no transcripts associated with this gene.
- Gene
- SP5
- Gene Name
- Sp5 transcription factor
View Full Product Details