Product Details

SNP ID
rs200252925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:166198967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACGCTGAATGACAGTAGCAGAC[A/G]CATCCTCTTGTTTCCGTTTTAGTGT
Phenotype
MIM: 603415
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929680 PubMed Links

Gene Details

Gene
LOC101929680
Gene Name
uncharacterized LOC101929680
There are no transcripts associated with this gene.

Gene
SCN9A
Gene Name
sodium voltage-gated channel alpha subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002977.3 5947 Missense Mutation GCG,GTG A1880V NP_002968.1
XM_005246757.2 5947 Missense Mutation GCG,GTG A1891V XP_005246814.1
XM_011511616.2 5947 Missense Mutation GCG,GTG A1891V XP_011509918.1
XM_011511617.2 5947 Missense Mutation GCG,GTG A1891V XP_011509919.1
XM_011511618.2 5947 Missense Mutation GCG,GTG A1880V XP_011509920.1
XM_011511619.2 5947 Intron XP_011509921.1
XM_017004668.1 5947 Missense Mutation GCG,GTG A1762V XP_016860157.1
XM_017004669.1 5947 Missense Mutation GCG,GTG A1643V XP_016860158.1

View Full Product Details