Product Details

SNP ID
rs199767522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241678591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCAGCTGTAACTGGAGGAACAGG[A/G]CCAGGTCGGGTTTGGGAAGGCCCAC
Phenotype
MIM: 611338 MIM: 188345 MIM: 608525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG4B PubMed Links

Gene Details

Gene
ATG4B
Gene Name
autophagy related 4B cysteine peptidase
There are no transcripts associated with this gene.

Gene
DTYMK
Gene Name
deoxythymidylate kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165031.1 517 Intron NP_001158503.1
NM_001320902.1 517 Missense Mutation GCC,GTC A87V NP_001307831.1
NM_001320903.1 517 Intron NP_001307832.1
NM_001320904.1 517 Missense Mutation GCC,GTC A111V NP_001307833.1
NM_001320905.1 517 Missense Mutation GCC,GTC A169V NP_001307834.1
NM_012145.3 517 Missense Mutation GCC,GTC A130V NP_036277.2
Gene
ING5
Gene Name
inhibitor of growth family member 5
There are no transcripts associated with this gene.

View Full Product Details