Product Details

SNP ID
rs199920386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69960771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCAGAAGGCGAAACTTCTTCCC[C/T]TTCAGGGTGCATGTGCGGTGCTCAA
Phenotype
MIM: 611765
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASPRV1 PubMed Links

Gene Details

Gene
ASPRV1
Gene Name
aspartic peptidase, retroviral-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152792.2 1495 Silent Mutation AAA,AAG K306K NP_690005.2
Gene
PCBP1-AS1
Gene Name
PCBP1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details