Product Details

SNP ID
rs199990516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:162144107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCAAGAATGGTGTTCATCTCATC[A/C]GAGAAAGAACCATCAGCATGTCTGC
Phenotype
MIM: 138030
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GCG PubMed Links

Gene Details

Gene
GCG
Gene Name
glucagon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002054.4 712 Silent Mutation NP_002045.1
Gene
LOC101929532
Gene Name
uncharacterized LOC101929532
There are no transcripts associated with this gene.

View Full Product Details