Product Details

SNP ID
rs200554227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:118942375 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAGCTGCTTTTCACCAAATTGC[A/G]ATGGAGCCTTTCGAAATCAATGGTA
Phenotype
MIM: 604870
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MARCO PubMed Links

Gene Details

Gene
MARCO
Gene Name
macrophage receptor with collagenous structure
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006770.3 210 Silent Mutation GCA,GCG A25A NP_006761.1
XM_011512082.1 210 Silent Mutation GCA,GCG A25A XP_011510384.1
XM_011512083.2 210 Silent Mutation GCA,GCG A25A XP_011510385.1
XM_017005171.1 210 Silent Mutation GCA,GCG A25A XP_016860660.1

View Full Product Details