Product Details

SNP ID
rs199713103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:136115220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGATGACTGTGGTCTTGAGGGC[C/T]TTGCGCTTCTGGTGGCCCTTGGAGT
Phenotype
MIM: 162643
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXCR4 PubMed Links

Gene Details

Gene
CXCR4
Gene Name
C-X-C motif chemokine receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008540.1 1024 Silent Mutation AAA,AAG K240K NP_001008540.1
NM_003467.2 1024 Silent Mutation AAA,AAG K236K NP_003458.1

View Full Product Details