Product Details

SNP ID
rs200426346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176080691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACGTGCGGGTAGTAGTGCAGCG[A/G]CACGTGCGAGTGGAAGGGGTAGGGC
Phenotype
MIM: 142991 MIM: 142989
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EVX2 PubMed Links

Gene Details

Gene
EVX2
Gene Name
even-skipped homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080458.1 1033 Missense Mutation CCG,TCG P283S NP_001073927.1
Gene
HOXD13
Gene Name
homeobox D13
There are no transcripts associated with this gene.

View Full Product Details