Product Details

SNP ID
rs199714967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:157415930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTCATCATCTGTACTCGTCTGC[C/T]GACTGAAGGCACCCCTGCTGGAGTC
Phenotype
MIM: 604448
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYTIP PubMed Links

Gene Details

Gene
CYTIP
Gene Name
cytohesin 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004288.4 1090 Missense Mutation CAG,CGG Q276R NP_004279.3
XM_017005386.1 1090 Missense Mutation CAG,CGG Q170R XP_016860875.1

View Full Product Details