Product Details

SNP ID
rs199783736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:55234159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGAGACTGATCTTTGCTGGCAA[A/G]CAGCTGGAAGATGGACGTACTTTGT
Phenotype
MIM: 603766 MIM: 191343
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLHC1 PubMed Links

Gene Details

Gene
CLHC1
Gene Name
clathrin heavy chain linker domain containing 1
There are no transcripts associated with this gene.

Gene
MTIF2
Gene Name
mitochondrial translational initiation factor 2
There are no transcripts associated with this gene.

Gene
RPS27A
Gene Name
ribosomal protein S27a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135592.2 367 Silent Mutation AAA,AAG K48K NP_001129064.1
NM_001177413.1 367 Silent Mutation AAA,AAG K48K NP_001170884.1
NM_002954.5 367 Silent Mutation AAA,AAG K48K NP_002945.1

View Full Product Details