Product Details

SNP ID
rs200132653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:227330691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCAGAAATTAGTCGAATTCAGT[A/G]CGAAATGGAATATACTGAAGGCATT
Phenotype
MIM: 614785
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC654841 PubMed Links

Gene Details

Gene
LOC654841
Gene Name
uncharacterized LOC654841
There are no transcripts associated with this gene.

Gene
MFF
Gene Name
mitochondrial fission factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277061.1 617 Intron NP_001263990.1
NM_001277062.1 617 Intron NP_001263991.1
NM_001277063.1 617 Intron NP_001263992.1
NM_001277064.1 617 Intron NP_001263993.1
NM_001277065.1 617 Intron NP_001263994.1
NM_001277066.1 617 Intron NP_001263995.1
NM_001277067.1 617 Intron NP_001263996.1
NM_001277068.1 617 Intron NP_001263997.1
NM_020194.5 617 Intron NP_064579.3
XM_005246681.3 617 Intron XP_005246738.1
XM_005246682.1 617 Intron XP_005246739.1
XM_005246683.1 617 Intron XP_005246740.1
XM_005246684.1 617 Intron XP_005246741.1
XM_005246685.3 617 Intron XP_005246742.1
XM_005246686.3 617 Intron XP_005246743.1
XM_005246687.3 617 Intron XP_005246744.1
XM_005246688.1 617 Intron XP_005246745.1
XM_006712637.1 617 Intron XP_006712700.1
XM_006712638.1 617 Intron XP_006712701.1
XM_006712639.2 617 Intron XP_006712702.1
XM_011511500.1 617 Intron XP_011509802.1
XM_011511501.1 617 Intron XP_011509803.1
XM_017004512.1 617 Missense Mutation TAC,TGC Y35C XP_016860001.1
XM_017004513.1 617 Intron XP_016860002.1
XM_017004514.1 617 Intron XP_016860003.1
XM_017004515.1 617 Intron XP_016860004.1

View Full Product Details