Product Details
- SNP ID
-
rs199980644
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:49922145 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGGAGGATAAGGATGCACAGGGC[A/G]GCAGCGGCTACTATCCCAACGACCA
- Phenotype
-
MIM: 600565
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NRXN1
PubMed Links
Gene Details
- Gene
- NRXN1
- Gene Name
- neurexin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
XM_005264642.3 |
5779 |
Silent Mutation |
GCC,GCT |
A1445A |
XP_005264699.1 |
XM_005264643.3 |
5779 |
Silent Mutation |
GCC,GCT |
A1430A |
XP_005264700.1 |
XM_006712137.3 |
5779 |
Silent Mutation |
GCC,GCT |
A1415A |
XP_006712200.1 |
XM_006712140.3 |
5779 |
Intron |
|
|
XP_006712203.2 |
XM_011533167.2 |
5779 |
Silent Mutation |
GCC,GCT |
A1448A |
XP_011531469.1 |
XM_011533171.2 |
5779 |
Silent Mutation |
GCC,GCT |
A1441A |
XP_011531473.1 |
XM_011533172.2 |
5779 |
Silent Mutation |
GCC,GCT |
A1439A |
XP_011531474.1 |
XM_011533174.2 |
5779 |
Silent Mutation |
GCC,GCT |
A1433A |
XP_011531476.1 |
XM_011533175.2 |
5779 |
Silent Mutation |
GCC,GCT |
A1429A |
XP_011531477.1 |
XM_011533177.2 |
5779 |
Silent Mutation |
GCC,GCT |
A1418A |
XP_011531479.1 |
XM_011533178.2 |
5779 |
Silent Mutation |
GCC,GCT |
A1418A |
XP_011531480.1 |
XM_011533180.2 |
5779 |
Intron |
|
|
XP_011531482.1 |
XM_011533183.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1159A |
XP_011531485.1 |
XM_017005303.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1457A |
XP_016860792.1 |
XM_017005304.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1456A |
XP_016860793.1 |
XM_017005305.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1454A |
XP_016860794.1 |
XM_017005306.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1453A |
XP_016860795.1 |
XM_017005307.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1451A |
XP_016860796.1 |
XM_017005308.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1450A |
XP_016860797.1 |
XM_017005309.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1448A |
XP_016860798.1 |
XM_017005310.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1447A |
XP_016860799.1 |
XM_017005311.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1442A |
XP_016860800.1 |
XM_017005312.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1440A |
XP_016860801.1 |
XM_017005313.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1438A |
XP_016860802.1 |
XM_017005314.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1437A |
XP_016860803.1 |
XM_017005315.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1436A |
XP_016860804.1 |
XM_017005316.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1436A |
XP_016860805.1 |
XM_017005317.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1434A |
XP_016860806.1 |
XM_017005318.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1433A |
XP_016860807.1 |
XM_017005319.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1432A |
XP_016860808.1 |
XM_017005320.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1429A |
XP_016860809.1 |
XM_017005321.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1427A |
XP_016860810.1 |
XM_017005322.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1424A |
XP_016860811.1 |
XM_017005323.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1419A |
XP_016860812.1 |
XM_017005324.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1409A |
XP_016860813.1 |
XM_017005325.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1406A |
XP_016860814.1 |
XM_017005326.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1405A |
XP_016860815.1 |
XM_017005327.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1400A |
XP_016860816.1 |
XM_017005328.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1394A |
XP_016860817.1 |
XM_017005329.1 |
5779 |
Intron |
|
|
XP_016860818.1 |
XM_017005330.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1386A |
XP_016860819.1 |
XM_017005331.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1381A |
XP_016860820.1 |
XM_017005332.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1374A |
XP_016860821.1 |
XM_017005333.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1371A |
XP_016860822.1 |
XM_017005334.1 |
5779 |
Silent Mutation |
GCC,GCT |
A1137A |
XP_016860823.1 |
XM_017005335.1 |
5779 |
Silent Mutation |
GCC,GCT |
A406A |
XP_016860824.1 |
XM_017005336.1 |
5779 |
Silent Mutation |
GCC,GCT |
A403A |
XP_016860825.1 |
XM_017005337.1 |
5779 |
Silent Mutation |
GCC,GCT |
A373A |
XP_016860826.1 |
View Full Product Details