Product Details

SNP ID
rs201823585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74461276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCTCCCCTCCTCTCCCTCCCTT[C/T]AGTAGTCTTCAGCCATGGCCAGTAA
Phenotype
MIM: 616456 MIM: 601336 MIM: 606961
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INO80B PubMed Links

Gene Details

Gene
INO80B
Gene Name
INO80 complex subunit B
There are no transcripts associated with this gene.

Gene
INO80B-WBP1
Gene Name
INO80B-WBP1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
MOGS
Gene Name
mannosyl-oligosaccharide glucosidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146158.1 2439 Silent Mutation TAA,TGA *732* NP_001139630.1
NM_006302.2 2439 Silent Mutation TAA,TGA *838* NP_006293.2
XM_017004876.1 2439 Silent Mutation TAA,TGA *563* XP_016860365.1
XM_017004877.1 2439 Silent Mutation TAA,TGA *563* XP_016860366.1
Gene
WBP1
Gene Name
WW domain binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012477.3 2439 Intron NP_036609.1

View Full Product Details