Product Details
- SNP ID
-
rs201587166
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:21002006 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGGCTTTTAATTATTTCCTGAGC[A/G]GTGGCAGAAAGCTCTGCAATCTTCT
- Phenotype
-
MIM: 107730
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
APOB
PubMed Links
Gene Details
- Gene
- APOB
- Gene Name
- apolipoprotein B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000384.2 |
13544 |
Silent Mutation |
ACC,ACT |
T4472T |
NP_000375.2 |
View Full Product Details