Product Details

SNP ID
rs202046520
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74135621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTTTGGGGACAGAGGTAAATATC[C/T]GCAATCCATGCATCTCTTTGATTTC
Phenotype
MIM: 613183
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
BOLA3 PubMed Links

Gene Details

Gene
BOLA3
Gene Name
bolA family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035505.1 Intron NP_001030582.1
NM_212552.2 Intron NP_997717.2

View Full Product Details