Product Details

SNP ID
rs201768943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:203706739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCTCAACTTATTCCCTTCAATT[A/C]AAGTAACAGGTAAACAATGTTAATG
Phenotype
MIM: 186760
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CD28 PubMed Links

Gene Details

Gene
CD28
Gene Name
CD28 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243077.1 265 Missense Mutation AAA,CAA K15Q NP_001230006.1
NM_001243078.1 265 Missense Mutation AAA,CAA K15Q NP_001230007.1
NM_006139.3 265 Missense Mutation AAA,CAA K15Q NP_006130.1
XM_011512194.2 265 Intron XP_011510496.1
XM_011512195.2 265 Intron XP_011510497.1
XM_011512197.2 265 Missense Mutation AAA,CAA K15Q XP_011510499.1

View Full Product Details