Product Details

SNP ID
rs202048301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:166198864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCATATCTTTTTTATTGAGTAA[A/G]TCATCATCTCTGTCTCCATCTTTTA
Phenotype
MIM: 603415
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929680 PubMed Links

Gene Details

Gene
LOC101929680
Gene Name
uncharacterized LOC101929680
There are no transcripts associated with this gene.

Gene
SCN9A
Gene Name
sodium voltage-gated channel alpha subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002977.3 6050 Silent Mutation GAC,GAT D1914D NP_002968.1
XM_005246757.2 6050 Silent Mutation GAC,GAT D1925D XP_005246814.1
XM_011511616.2 6050 Silent Mutation GAC,GAT D1925D XP_011509918.1
XM_011511617.2 6050 Silent Mutation GAC,GAT D1925D XP_011509919.1
XM_011511618.2 6050 Silent Mutation GAC,GAT D1914D XP_011509920.1
XM_011511619.2 6050 Intron XP_011509921.1
XM_017004668.1 6050 Silent Mutation GAC,GAT D1796D XP_016860157.1
XM_017004669.1 6050 Silent Mutation GAC,GAT D1677D XP_016860158.1

View Full Product Details