Product Details

SNP ID
rs201520712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197540123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCATTTTCAGGCAGTTCTGCCT[C/T]GAGCTAAATGGACTTGCTGTCAAAC
Phenotype
MIM: 609361
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSPE1-MOB4 PubMed Links

Gene Details

Gene
HSPE1-MOB4
Gene Name
HSPE1-MOB4 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202485.1 235 Silent Mutation CTC,CTT L115L NP_001189414.1
Gene
MOB4
Gene Name
MOB family member 4, phocein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100819.2 235 Silent Mutation CTC,CTT L58L NP_001094289.1
NM_001204094.1 235 Silent Mutation CTC,CTT L47L NP_001191023.1
NM_015387.4 235 Silent Mutation CTC,CTT L79L NP_056202.2
NM_199482.3 235 Silent Mutation CTC,CTT L47L NP_955776.1

View Full Product Details