Product Details

SNP ID
rs201882166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101008047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAATGCCTGGGAGTCCTGGGGC[A/G]TCTTCCCAGTGTCTGCAAAAACCGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL31 PubMed Links

Gene Details

Gene
RPL31
Gene Name
ribosomal protein L31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000993.4 3617 Intron NP_000984.1
NM_001098577.2 3617 Intron NP_001092047.1
NM_001099693.1 3617 Intron NP_001093163.1
Gene
TBC1D8
Gene Name
TBC1 domain family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005263862.3 3617 Missense Mutation XP_005263919.1
XM_005263863.4 3617 Missense Mutation XP_005263920.1
XM_005263864.3 3617 Missense Mutation XP_005263921.1
XM_011510507.1 3617 Missense Mutation XP_011508809.1
XM_011510508.2 3617 Intron XP_011508810.1
XM_017003198.1 3617 Missense Mutation XP_016858687.1
XM_017003199.1 3617 Missense Mutation XP_016858688.1
XM_017003200.1 3617 Intron XP_016858689.1
XM_017003201.1 3617 Intron XP_016858690.1
XM_017003202.1 3617 Intron XP_016858691.1

View Full Product Details