Product Details

SNP ID
rs202188020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:181457736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGATGCTGTTGCTGTGCCTGGGG[G/T]TCCCGACCGGCCGCCCCTACAACGT
Phenotype
MIM: 192975
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ITGA4 PubMed Links

Gene Details

Gene
ITGA4
Gene Name
integrin subunit alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000885.5 845 Missense Mutation GTC,TTC V28F NP_000876.3
NM_001316312.1 845 Missense Mutation GTC,TTC V28F NP_001303241.1

View Full Product Details