Product Details

SNP ID
rs201925005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:230037513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGAAGGAAAAATCATATTTTTGC[A/G]TGATGTCATAGATCCACCCTACAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC16A14 PubMed Links

Gene Details

Gene
SLC16A14
Gene Name
solute carrier family 16 member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152527.4 1715 Missense Mutation ACG,ATG T467M NP_689740.2
XM_005246353.3 1715 Missense Mutation ACG,ATG T419M XP_005246410.1
XM_011510751.2 1715 Missense Mutation ACG,ATG T467M XP_011509053.1
XM_011510752.1 1715 Intron XP_011509054.1
XM_017003480.1 1715 Missense Mutation ACG,ATG T467M XP_016858969.1

View Full Product Details