Product Details

SNP ID
rs202107509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:11658596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCTCTCCACACAGGGAGCTGAC[A/G]GCTTCCAGGAAGAGTTTTCTGAAGG
Phenotype
MIM: 605538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NTSR2 PubMed Links

Gene Details

Gene
NTSR2
Gene Name
neurotensin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012344.3 1190 Silent Mutation GCC,GCT A372A NP_036476.1
XM_005246156.3 1190 UTR 3 XP_005246213.1
XM_006711876.3 1190 UTR 3 XP_006711939.1
XM_006711877.3 1190 UTR 3 XP_006711940.1
XM_017003738.1 1190 UTR 3 XP_016859227.1

View Full Product Details