Product Details

SNP ID
rs202122063
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:20667573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGCCGGGGCAGCCGCTGTCTCC[A/G]CCTCGGGCCATGGTCGCGCGGACAC
Phenotype
MIM: 604651
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GDF7 PubMed Links
Additional Information
For this assay, SNP(s) [rs140362230] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GDF7
Gene Name
growth differentiation factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182828.3 910 Missense Mutation ACC,GCC T112A NP_878248.2

View Full Product Details