Product Details

SNP ID
rs201898015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241603061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGTGCATCGGCTTGCGCGTGTC[C/T]GGGTGGAAGGAGTTGAACCTGGACG
Phenotype
MIM: 612533
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
THAP4 PubMed Links

Gene Details

Gene
THAP4
Gene Name
THAP domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164356.1 1712 Silent Mutation CCA,CCG P61P NP_001157828.1
NM_015963.5 1712 Silent Mutation CCA,CCG P473P NP_057047.4
XM_005247016.4 1712 Silent Mutation CCA,CCG P580P XP_005247073.3
XM_011511291.2 1712 Silent Mutation CCA,CCG P497P XP_011509593.1
XM_017004256.1 1712 Silent Mutation CCA,CCG P497P XP_016859745.1

View Full Product Details