Product Details

SNP ID
rs202039098
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37171452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTACCTTTGCGGAAAAGGAATG[G/T]GCCGACAGCACCGTGTGTGTCCTGA
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
SULT6B1 PubMed Links

Gene Details

Gene
SULT6B1
Gene Name
sulfotransferase family 6B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032377.1 670 Missense Mutation ACA,CCA T217P NP_001027549.1

View Full Product Details