Product Details
- SNP ID
-
rs202245854
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:54973169 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTGGGCGTTTTCATTCAGCTTTG[C/T]GCTTCAATCCAGGGATTTTTGCTTG
- Phenotype
-
MIM: 604475
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EML6
PubMed Links
Gene Details
- Gene
- EML6
- Gene Name
- echinoderm microtubule associated protein like 6
- Gene
- RTN4
- Gene Name
- reticulon 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001321859.1 |
801 |
Missense Mutation |
CAC,CGC |
H983R |
NP_001308788.1 |
NM_001321860.1 |
801 |
Missense Mutation |
CAC,CGC |
H983R |
NP_001308789.1 |
NM_001321861.1 |
801 |
Missense Mutation |
CAC,CGC |
H983R |
NP_001308790.1 |
NM_001321862.1 |
801 |
Missense Mutation |
CAC,CGC |
H983R |
NP_001308791.1 |
NM_001321863.1 |
801 |
Missense Mutation |
CAC,CGC |
H983R |
NP_001308792.1 |
NM_001321904.1 |
801 |
Missense Mutation |
CAC,CGC |
H983R |
NP_001308833.1 |
NM_007008.2 |
801 |
Missense Mutation |
CAC,CGC |
H196R |
NP_008939.1 |
NM_020532.4 |
801 |
Missense Mutation |
CAC,CGC |
H1189R |
NP_065393.1 |
NM_153828.2 |
801 |
Missense Mutation |
CAC,CGC |
H370R |
NP_722550.1 |
NM_207520.1 |
801 |
Missense Mutation |
CAC,CGC |
H389R |
NP_997403.1 |
NM_207521.1 |
801 |
Missense Mutation |
CAC,CGC |
H983R |
NP_997404.1 |
XM_005264434.3 |
801 |
Missense Mutation |
CAC,CGC |
H983R |
XP_005264491.1 |
XM_017004518.1 |
801 |
Missense Mutation |
CAC,CGC |
H983R |
XP_016860007.1 |
XM_017004519.1 |
801 |
Missense Mutation |
CAC,CGC |
H983R |
XP_016860008.1 |
View Full Product Details