Product Details

SNP ID
rs201470529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:160278610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCCAGAGGGCTGATCATTGATG[C/T]GGGCTGTAGTGACATGGTGTGCTCC
Phenotype
MIM: 602310
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100505984 PubMed Links

Gene Details

Gene
LOC100505984
Gene Name
uncharacterized LOC100505984
There are no transcripts associated with this gene.

Gene
RBMS1
Gene Name
RNA binding motif single stranded interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002897.4 1408 Missense Mutation ACA,GCA T331A NP_002888.1
NM_016836.3 1408 Missense Mutation ACA,GCA T334A NP_058520.1
XM_005246737.1 1408 Missense Mutation ACA,GCA T350A XP_005246794.1
XM_005246738.2 1408 Missense Mutation ACA,GCA T349A XP_005246795.1
XM_005246739.2 1408 Missense Mutation ACA,GCA T348A XP_005246796.1
XM_005246740.1 1408 Missense Mutation ACA,GCA T347A XP_005246797.1
XM_005246741.3 1408 Missense Mutation ACA,GCA T347A XP_005246798.1
XM_017004626.1 1408 Missense Mutation ACA,GCA T330A XP_016860115.1

View Full Product Details