Product Details

SNP ID
rs201944648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:79085578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGACTTGTAGGAGACCAGGGAC[C/T]CACTACTCCAGTGCCAGCGGCGGTT
Phenotype
MIM: 167771
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
REG1B PubMed Links

Gene Details

Gene
REG1B
Gene Name
regenerating family member 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006507.3 459 Missense Mutation GAG,GGG E116G NP_006498.1

View Full Product Details