Product Details

SNP ID
rs201711130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74529161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACAGGTGGTAAGCAAATTGACTA[C/T]GTTGTGGTCGAAAGGTGTCACATGG
Phenotype
MIM: 602434 MIM: 606441 MIM: 607163
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AUP1 PubMed Links

Gene Details

Gene
AUP1
Gene Name
ancient ubiquitous protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181575.4 523 Missense Mutation ATA,GTA I104V NP_853553.1
XM_005264392.3 523 Missense Mutation ATA,GTA I161V XP_005264449.1
Gene
DQX1
Gene Name
DEAQ-box RNA dependent ATPase 1
There are no transcripts associated with this gene.

Gene
HTRA2
Gene Name
HtrA serine peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321727.1 523 Intron NP_001308656.1
NM_001321728.1 523 Intron NP_001308657.1
NM_013247.4 523 Intron NP_037379.1
NM_145074.2 523 Intron NP_659540.1
Gene
LOXL3
Gene Name
lysyl oxidase like 3
There are no transcripts associated with this gene.

View Full Product Details